siRNA Details
| Gene Symbol: LOC100911140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771277.1 | XP_008769499.1 | 4 | 2278 | ||
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771274.1 | XP_008769496.1 | 2360 | |||
| GenBank mRNA | BC166517.1 | - | 2360 | 7132003 | ||

LOC100911140
Genomic Map