siRNA Details
| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106085.1 | NP_001099555.1 | 7 | 598 | ||
| GenBank mRNA | FQ212035.1 | - | 7 | 584 | ||
| FQ226689.1 | - | 7 | 621 | |||
| FQ233791.1 | - | 7 | 615 | |||
| FQ234632.1 | - | 7 | 621 | |||

Dctn6
Genomic Map