siRNA Details
| Gene Symbol: Whsc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106090.1 | NP_001099560.1 | 5 | 1434 | ||
| XM_006253307.2 | XP_006253369.1 | 5 | 1551 | |||
| XM_006253309.2 | XP_006253371.1 | 5 | 1551 | |||
| XM_006253311.2 | XP_006253373.1 | 5 | 1551 | |||
| XM_008771349.1 | XP_008769571.1 | 5 | 3422 | |||
| XM_008771350.1 | XP_008769572.1 | 5 | 1550 | |||

Whsc1l1
Genomic Map