siRNA Details
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106092.2 | NP_001099562.2 | 10, 11 | 874 | ||
GenBank mRNA | BC160860.1 | - | 10, 11 | 874 | 7321877 | |
FQ217181.1 | - | 10, 11 | 885 |
