siRNA Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106093.2 | NP_001099563.1 | 17 | 2222 | ||
| NM_001135012.1 | NP_001128484.1 | 18 | 2363 | |||
| GenBank mRNA | BC166460.1 | - | 18 | 2361 | 9027936 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp4| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106093.2 | NP_001099563.1 | 17 | 2222 | ||
| NM_001135012.1 | NP_001128484.1 | 18 | 2363 | |||
| GenBank mRNA | BC166460.1 | - | 18 | 2361 | 9027936 | |