siRNA Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106093.2 | NP_001099563.1 | 10, 11 | 1383 | ||
| NM_001135012.1 | NP_001128484.1 | 11, 12 | 1524 | |||
| GenBank mRNA | BC166460.1 | - | 11, 12 | 1522 | 9027936 | |

Usp4
Genomic Map