siRNA Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106094.2 | NP_001099564.1 | 1 | 65 | ||
| XM_008771387.1 | XP_008769609.1 | 7 | 813 | |||
| XM_008771388.1 | XP_008769610.1 | 6 | 752 | |||
| XM_008771389.1 | XP_008769611.1 | 6 | 724 | |||
| GenBank mRNA | BC169123.1 | - | 1 | 65 | 6887046 | |

Tmem255b
Genomic Map