siRNA Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771387.1 | XP_008769609.1 | 6 | 654 | ||
| XM_008771388.1 | XP_008769610.1 | 5 | 593 | |||
| XM_008771389.1 | XP_008769611.1 | 5 | 565 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem255b| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771387.1 | XP_008769609.1 | 6 | 654 | ||
| XM_008771388.1 | XP_008769610.1 | 5 | 593 | |||
| XM_008771389.1 | XP_008769611.1 | 5 | 565 | |||