siRNA Details
| Gene Symbol: Col4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135009.1 | NP_001128481.1 | 51 | 5113 | ||
| GenBank mRNA | BC089096.1 | - | 5 | 694 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col4a1| Gene Symbol: Col4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135009.1 | NP_001128481.1 | 51 | 5113 | ||
| GenBank mRNA | BC089096.1 | - | 5 | 694 | ||