siRNA Details
| Gene Symbol: Aga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031641.1 | NP_001026811.1 | 3 | 348 | ||
| XM_006253111.2 | XP_006253173.1 | 4 | 473 | |||
| GenBank mRNA | BC098718.1 | - | 3 | 348 | 7366394 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aga| Gene Symbol: Aga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031641.1 | NP_001026811.1 | 3 | 348 | ||
| XM_006253111.2 | XP_006253173.1 | 4 | 473 | |||
| GenBank mRNA | BC098718.1 | - | 3 | 348 | 7366394 | |