siRNA Details
| Gene Symbol: Npepo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012346.1 | NP_001012346.1 | 1 | 292 | ||
| XM_008771429.1 | XP_008769651.1 | 3 | 740 | |||
| XM_008771430.1 | XP_008769652.1 | 3 | 740 | |||
| XM_008771431.1 | XP_008769653.1 | 2 | 587 | |||
| XM_008771432.1 | XP_008769654.1 | 2 | 579 | |||
| XM_008771433.1 | XP_008769655.1 | 2 | 452 | |||
| GenBank mRNA | AJ810421.1 | - | 1 | 292 | ||

Npepo
Genomic Map