siRNA Details
Gene Symbol: Catsper3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106101.1 | NP_001099571.1 | 7 | 1041 | ||
XM_006253572.2 | XP_006253634.1 | 7 | 2276 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Catsper3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106101.1 | NP_001099571.1 | 7 | 1041 | ||
XM_006253572.2 | XP_006253634.1 | 7 | 2276 |