siRNA Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011922.1 | NP_001011922.1 | 5 | 1089 | ||
| XM_006253786.2 | XP_006253848.1 | 5 | 1252 | |||
| XM_008771569.1 | XP_008769791.1 | 5 | 1905 | |||
| GenBank mRNA | BC085812.1 | - | 5 | 1089 | 7121465 | |

Nedd9
Genomic Map