siRNA Details
| Gene Symbol: Rpp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013055.1 | NP_001013073.1 | 3, 4 | 398 | ||
| XM_006253790.1 | XP_006253852.1 | 3, 4 | 360 | |||
| GenBank mRNA | BC091191.1 | - | 3, 4 | 398 | 7368273 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpp40| Gene Symbol: Rpp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013055.1 | NP_001013073.1 | 3, 4 | 398 | ||
| XM_006253790.1 | XP_006253852.1 | 3, 4 | 360 | |||
| GenBank mRNA | BC091191.1 | - | 3, 4 | 398 | 7368273 | |