siRNA Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006966.1 | NP_001006967.1 | 10 | 1079 | ||
| GenBank mRNA | BC083764.1 | - | 10 | 1079 | 7193133 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eci2| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006966.1 | NP_001006967.1 | 10 | 1079 | ||
| GenBank mRNA | BC083764.1 | - | 10 | 1079 | 7193133 | |