siRNA Details
| Gene Symbol: Dcdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106110.2 | NP_001099580.2 | 10 | 1440 | ||
| GenBank mRNA | FM100065.1 | - | 4 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcdc2| Gene Symbol: Dcdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106110.2 | NP_001099580.2 | 10 | 1440 | ||
| GenBank mRNA | FM100065.1 | - | 4 | 550 | ||