siRNA Details
| Gene Symbol: Trim27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134974.1 | NP_001128446.1 | 8 | 1771 | ||
| GenBank mRNA | BC169072.1 | - | 8 | 1771 | 5599836 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim27| Gene Symbol: Trim27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134974.1 | NP_001128446.1 | 8 | 1771 | ||
| GenBank mRNA | BC169072.1 | - | 8 | 1771 | 5599836 | |