siRNA Details
| Gene Symbol: Actn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170325.1 | NP_001163796.1 | 19, 20 | 2566 | ||
| GenBank mRNA | FM118105.1 | - | 4, 5 | 481 | ||
| FM123616.1 | - | 3, 4 | 280 | |||
| FQ224888.1 | - | 5, 6 | 705 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actn2| Gene Symbol: Actn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170325.1 | NP_001163796.1 | 19, 20 | 2566 | ||
| GenBank mRNA | FM118105.1 | - | 4, 5 | 481 | ||
| FM123616.1 | - | 3, 4 | 280 | |||
| FQ224888.1 | - | 5, 6 | 705 | |||