siRNA Details
| Gene Symbol: Tubal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106118.1 | NP_001099588.1 | 3 | 443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubal3| Gene Symbol: Tubal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106118.1 | NP_001099588.1 | 3 | 443 | ||