siRNA Details
| Gene Symbol: Rpp38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033063.1 | NP_001028235.1 | 3 | 745 | ||
| XM_006254267.2 | XP_006254329.1 | 2 | 1204 | |||
| XM_006254268.2 | XP_006254330.1 | 2 | 724 | |||
| GenBank mRNA | BC100623.1 | - | 3 | 745 | 7929545 | |

Rpp38
Genomic Map