siRNA Details
| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106123.2 | NP_001099593.2 | 18 | 2703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrc1| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106123.2 | NP_001099593.2 | 18 | 2703 | ||