siRNA Details
| Gene Symbol: Atp1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271030.1 | NP_001257959.1 | 10 | 1422 | ||
| NM_022848.3 | NP_074039.1 | 10 | 1425 | |||
| GenBank mRNA | BC087015.1 | - | 10 | 1414 | 7113151 | |
| U15176.1 | - | 10 | 1425 | |||

Atp1a4
Genomic Map