siRNA Details
| Gene Symbol: Prtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106127.1 | NP_001099597.1 | 1, 2 | 83 | ||
| XM_008771860.1 | XP_008770082.1 | 1, 2 | 94 | |||
| GenBank mRNA | BC166887.1 | - | 1, 2 | 80 | 6890546 | |
| BC166923.1 | - | 1, 2 | 80 | 6919422 | ||

Prtfdc1
Genomic Map