siRNA Details
| Gene Symbol: Hpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017112.1 | NP_058808.1 | 10 | 1177 | ||
| GenBank mRNA | FQ219040.1 | - | 10 | 1075 | ||
| X70900.1 | - | 10 | 1177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpn| Gene Symbol: Hpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017112.1 | NP_058808.1 | 10 | 1177 | ||
| GenBank mRNA | FQ219040.1 | - | 10 | 1075 | ||
| X70900.1 | - | 10 | 1177 | |||