siRNA Details
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728514.4 | XP_002728560.1 | 1 | 490 | ||
GenBank mRNA | FQ231772.1 | - | 1 | 483 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728514.4 | XP_002728560.1 | 1 | 490 | ||
GenBank mRNA | FQ231772.1 | - | 1 | 483 |