siRNA Details
| Gene Symbol: Dnajc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728514.4 | XP_002728560.1 | 4 | 766 | ||
| GenBank mRNA | FQ231772.1 | - | 4 | 759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc1| Gene Symbol: Dnajc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728514.4 | XP_002728560.1 | 4 | 766 | ||
| GenBank mRNA | FQ231772.1 | - | 4 | 759 | ||