siRNA Details
| Gene Symbol: Cbln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012740.1 | NP_001012758.1 | 2, 3 | 664 | ||
| XM_006255022.2 | XP_006255084.1 | 4, 5 | 1165 | |||
| XM_006255023.2 | XP_006255085.1 | 3, 4 | 1120 | |||
| XM_006255024.2 | XP_006255086.1 | 3, 4 | 800 | |||
| GenBank mRNA | BC091341.1 | - | 2, 3 | 664 | 7315458 | |

Cbln2
Genomic Map