siRNA Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106133.2 | NP_001099603.2 | 11, 12 | 1506 | ||
| XM_008772103.1 | XP_008770325.1 | 11, 12 | 1571 | |||
| XM_008772104.1 | XP_008770326.1 | 11, 12 | 1571 | |||
| GenBank mRNA | BC168151.1 | - | 11, 12 | 1500 | 7459379 | |
| DV722630.1 | - | 1, 2 | 70 | |||

Dym
Genomic Map