siRNA Details
| Gene Symbol: Mbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011924.1 | NP_001011924.1 | 10 | 1094 | ||
| XM_006254909.2 | XP_006254971.1 | 12 | 1536 | |||
| XM_006254910.2 | XP_006254972.1 | 12 | 1533 | |||
| XM_006254911.2 | XP_006254973.1 | 12 | 1536 | |||
| XM_006254913.2 | XP_006254975.1 | 11 | 1367 | |||
| XM_006254914.2 | XP_006254976.1 | 11 | 1367 | |||
| XM_008772105.1 | XP_008770327.1 | 13 | 1613 | |||
| GenBank mRNA | BC081932.1 | - | 10 | 1094 | 7114194 | |

Mbd1
Genomic Map