siRNA Details
| Gene Symbol: Canx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172008.2 | NP_742005.1 | 6, 7 | 628 | ||
| XM_008767679.1 | XP_008765901.1 | 6, 7 | 749 | |||
| XM_008767680.1 | XP_008765902.1 | 6, 7 | 668 | |||
| GenBank mRNA | EV776124.1 | - | 4, 5 | 357 | ||
| FQ210638.1 | - | 6, 7 | 654 | |||
| L18889.1 | - | 5, 6 | 525 | |||

Canx
Genomic Map