siRNA Details
| Gene Symbol: Canx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172008.2 | NP_742005.1 | 2 | 143 | ||
| XM_008767679.1 | XP_008765901.1 | 2 | 264 | |||
| XM_008767680.1 | XP_008765902.1 | 2 | 183 | |||
| GenBank mRNA | CB586187.1 | - | 2 | 143 | ||
| FQ210638.1 | - | 2 | 169 | |||
| L18889.1 | - | 1 | 40 | |||

Canx
Genomic Map