siRNA Details
| Gene Symbol: Srfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005536.1 | NP_001005536.1 | 6 | 490 | ||
| XM_008772109.1 | XP_008770331.1 | 4 | 371 | |||
| GenBank mRNA | BC082077.1 | - | 6 | 490 | 7133229 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srfbp1| Gene Symbol: Srfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005536.1 | NP_001005536.1 | 6 | 490 | ||
| XM_008772109.1 | XP_008770331.1 | 4 | 371 | |||
| GenBank mRNA | BC082077.1 | - | 6 | 490 | 7133229 | |