siRNA Details
| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240666.2 | XP_006240728.1 | 12 | 1747 | ||
| XM_006240667.2 | XP_006240729.1 | 11 | 1632 | |||
| GenBank mRNA | U70050.1 | - | 12 | 1562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jag2| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240666.2 | XP_006240728.1 | 12 | 1747 | ||
| XM_006240667.2 | XP_006240729.1 | 11 | 1632 | |||
| GenBank mRNA | U70050.1 | - | 12 | 1562 | ||