siRNA Details
| Gene Symbol: Mstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019151.1 | NP_062024.1 | 1 | 321 | ||
| GenBank mRNA | AF019624.1 | - | 1 | 321 | ||
| FQ224469.1 | - | 445 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mstn| Gene Symbol: Mstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019151.1 | NP_062024.1 | 1 | 321 | ||
| GenBank mRNA | AF019624.1 | - | 1 | 321 | ||
| FQ224469.1 | - | 445 | ||||