siRNA Details
| Gene Symbol: Psmg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106138.2 | NP_001099608.2 | 6 | 759 | ||
| GenBank mRNA | BM387084.1 | - | 1, 2 | 319 | ||
| FQ212569.1 | - | 6 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmg2| Gene Symbol: Psmg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106138.2 | NP_001099608.2 | 6 | 759 | ||
| GenBank mRNA | BM387084.1 | - | 1, 2 | 319 | ||
| FQ212569.1 | - | 6 | 823 | |||