siRNA Details
| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100514.1 | NP_001093984.1 | 7 | 1067 | ||
| XM_006254858.2 | XP_006254920.1 | 7 | 1358 | |||
| XM_008772118.1 | XP_008770340.1 | 6 | 761 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep76| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100514.1 | NP_001093984.1 | 7 | 1067 | ||
| XM_006254858.2 | XP_006254920.1 | 7 | 1358 | |||
| XM_008772118.1 | XP_008770340.1 | 6 | 761 | |||