siRNA Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017116.2 | NP_058812.1 | 20 | 2473 | ||
| GenBank mRNA | BC065306.1 | - | 20 | 2473 | 5621252 | |
| FQ209553.1 | - | 1 | 24 | |||
| FQ225985.1 | - | 1 | 28 | |||
| L09120.1 | - | 20 | 2159 | |||

Capn2
Genomic Map