siRNA Details
| Gene Symbol: Nedd4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008300.2 | NP_001008301.2 | 25 | 2448 | ||
| GenBank mRNA | BC061798.1 | - | 4 | 274 | ||
| BC086371.1 | - | 9 | 970 | |||
| GQ160815.1 | - | 24 | 2388 | |||
| GQ160816.1 | - | 24 | 2388 | |||

Nedd4l
Genomic Map