siRNA Details
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008300.2 | NP_001008301.2 | 15 | 1387 | ||
GenBank mRNA | GQ160815.1 | - | 14 | 1327 | ||
GQ160816.1 | - | 14 | 1327 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008300.2 | NP_001008301.2 | 15 | 1387 | ||
GenBank mRNA | GQ160815.1 | - | 14 | 1327 | ||
GQ160816.1 | - | 14 | 1327 |