siRNA Details
| Gene Symbol: Nars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025635.2 | NP_001020806.2 | 15 | 1645 | ||
| NM_001142949.1 | NP_001136421.1 | 13 | 1492 | |||
| XM_006254844.2 | XP_006254906.1 | 15 | 1703 | |||
| GenBank mRNA | AY387099.1 | - | 12 | 1398 | ||
| BC099099.1 | - | 14 | 1526 | 7459693 | ||

Nars
Genomic Map