siRNA Details
| Gene Symbol: LOC100911363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748836.3 | XP_003748884.1 | 1, 2 | 92 | ||
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017118.1 | NP_058814.1 | 1, 2 | 43 | ||
| GenBank mRNA | BC098068.1 | - | 1, 2 | 43 | 7462537 | |

LOC100911363
Genomic Map