siRNA Details
| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106143.1 | NP_001099613.1 | 4 | 468 | ||
| XM_008772124.1 | XP_008770346.1 | 4 | 451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcof1| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106143.1 | NP_001099613.1 | 4 | 468 | ||
| XM_008772124.1 | XP_008770346.1 | 4 | 451 | |||