siRNA Details
Gene Symbol: Ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105725.2 | NP_001099195.1 | 3 | 444 | ||
GenBank mRNA | BC169085.1 | - | 3 | 444 | 7938657 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105725.2 | NP_001099195.1 | 3 | 444 | ||
GenBank mRNA | BC169085.1 | - | 3 | 444 | 7938657 |