siRNA Details
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106148.1 | NP_001099618.1 | 2 | 586 | ||
XM_006254666.2 | XP_006254728.1 | 2 | 483 | |||
XM_006254667.2 | XP_006254729.1 | 2 | 600 | |||
XM_006254668.2 | XP_006254730.1 | 2 | 483 |
Gene Symbol: LOC100909761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254990.2 | XP_006255052.1 | 2 | 495 | ||
XM_006254991.2 | XP_006255053.1 | 2 | 612 | |||
XM_006254992.2 | XP_006255054.1 | 2 | 495 |
Gene Symbol: LOC100912306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244679.2 | XP_006244741.1 | 2 | 483 | ||
XM_006244680.2 | XP_006244742.1 | 2 | 600 | |||
XM_006244681.2 | XP_006244743.1 | 2 | 483 |
