siRNA Details
| Gene Symbol: Cdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012737.1 | NP_001012755.1 | 11 | 2029 | ||
| GenBank mRNA | AB121031.1 | - | 11 | 2029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh7| Gene Symbol: Cdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012737.1 | NP_001012755.1 | 11 | 2029 | ||
| GenBank mRNA | AB121031.1 | - | 11 | 2029 | ||