siRNA Details
| Gene Symbol: Cdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168630.1 | NP_001162101.1 | 6 | 1235 | ||
| XM_006232076.2 | XP_006232138.1 | 8 | 1459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh9| Gene Symbol: Cdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168630.1 | NP_001162101.1 | 6 | 1235 | ||
| XM_006232076.2 | XP_006232138.1 | 8 | 1459 | |||