siRNA Details
| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106154.1 | NP_001099624.1 | 7 | 918 | ||
| GenBank mRNA | BC166835.1 | - | 7 | 780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco6| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106154.1 | NP_001099624.1 | 7 | 918 | ||
| GenBank mRNA | BC166835.1 | - | 7 | 780 | ||