siRNA Details
Gene Symbol: Hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100658.2 | NP_001094128.2 | 3 | 273 | ||
GenBank mRNA | S75280.1 | - | 3 | 220 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100658.2 | NP_001094128.2 | 3 | 273 | ||
GenBank mRNA | S75280.1 | - | 3 | 220 |