siRNA Details
| Gene Symbol: Fam13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106158.1 | NP_001099628.1 | 11, 12 | 1743 | ||
| XM_008772033.1 | XP_008770255.1 | 10, 11 | 1367 | |||
| GenBank mRNA | FQ228342.1 | - | 9, 10 | 1255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam13b| Gene Symbol: Fam13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106158.1 | NP_001099628.1 | 11, 12 | 1743 | ||
| XM_008772033.1 | XP_008770255.1 | 10, 11 | 1367 | |||
| GenBank mRNA | FQ228342.1 | - | 9, 10 | 1255 | ||