siRNA Details
| Gene Symbol: Slc39a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024745.1 | NP_001019916.1 | 5 | 1454 | ||
| GenBank mRNA | BC097493.1 | - | 5 | 1454 | 7461645 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a6| Gene Symbol: Slc39a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024745.1 | NP_001019916.1 | 5 | 1454 | ||
| GenBank mRNA | BC097493.1 | - | 5 | 1454 | 7461645 | |